Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc144b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178418.4 | NP_848505.2 | 2, 3 | 258 | ||
GenBank mRNA | AK133178.1 | - | 2, 3 | 244 | ||
BC050811.1 | - | 2, 3 | 260 | 6772771 |