Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppm1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178726.3 | NP_848841.2 | 3, 4 | 1390 | ||
GenBank mRNA | AF117832.1 | - | 2, 3 | 345 | ||
AK028275.1 | - | 3, 4 | 432 | |||
AK032529.1 | - | 3, 4 | 1448 | |||
AK035912.1 | - | 3, 4 | 1393 | |||
AK045724.1 | - | 3, 4 | 1431 | |||
AK131646.1 | - | 3, 4 | 1377 | |||
AK142789.1 | - | 3, 4 | 1409 | |||
AK147389.1 | - | 3, 4 | 1391 | |||
AK147876.1 | - | 3, 4 | 1427 | |||
AK165160.1 | - | 3, 4 | 1423 | |||
AK191249.1 | - | 1, 2 | 115 | |||
AK220523.1 | - | 3, 4 | 1395 | |||
AY184801.1 | - | 3, 4 | 978 | |||
BC096031.1 | - | 3, 4 | 1390 | 6837066 |