Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172864.3 | NP_766452.2 | 20, 21 | 2553 | ||
GenBank mRNA | AK029903.1 | - | 20, 21 | 2555 | ||
BC132106.1 | - | 15, 16 | 1991 | 40130383 | ||
BC132108.1 | - | 15, 16 | 1991 | 40130385 | ||
BC158119.1 | - | 20, 21 | 2659 | 9007392 |