Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172866.3 | NP_766454.1 | 5, 6 | 774 | ||
GenBank mRNA | AK089288.1 | - | 5, 6 | 557 | ||
AK129103.1 | - | 5, 6 | 560 | |||
AK170117.1 | - | 5, 6 | 571 | |||
BC086614.1 | - | 5, 6 | 580 | 30533256 |