Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172866.3 | NP_766454.1 | 9 | 1290 | ||
GenBank mRNA | AK089288.1 | - | 9 | 1073 | ||
AK129103.1 | - | 9 | 1076 | |||
AK170117.1 | - | 9 | 1087 | |||
BC086614.1 | - | 9 | 1096 | 30533256 |