Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Palm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172868.3 | NP_766456.1 | 3 | 333 | ||
GenBank mRNA | AK045344.1 | - | 3 | 334 | ||
AK133666.1 | - | 294 | ||||
BC113156.1 | - | 3 | 299 | 40058559 |
Gene Symbol: Pakap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BF469297.1 | - | 5 | 336 |