Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Palm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172868.3 | NP_766456.1 | 2, 3 | 241 | ||
GenBank mRNA | AK045344.1 | - | 2, 3 | 242 | ||
AK133666.1 | - | 202 | ||||
BC113156.1 | - | 2, 3 | 207 | 40058559 |