Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Raver2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183024.1 | NP_898845.1 | 7, 8 | 1333 | ||
GenBank mRNA | AK029114.1 | - | 5, 6 | 829 | ||
AK040404.1 | - | 7, 8 | 1390 | |||
BC055329.1 | - | 7, 8 | 1333 | 6835616 |