Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Raver2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183024.1 | NP_898845.1 | 6 | 1207 | ||
GenBank mRNA | AK029114.1 | - | 4 | 703 | ||
AK040404.1 | - | 6 | 1264 | |||
BC055329.1 | - | 6 | 1207 | 6835616 |