Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlgap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK158868.1 | - | 5, 6 | 1990 | ||
AY243848.2 | - | 3, 4 | 1370 | |||
BC057615.1 | - | 5, 6 | 1599 | 6401651 | ||
BC058433.1 | - | 3, 4 | 797 | |||
BC098311.1 | - | 2132 |