Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rundc3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198620.1 | NP_941022.1 | 10 | 1468 | ||
GenBank mRNA | BC057375.1 | - | 6 | 681 | ||
BC058808.1 | - | 10 | 1468 | 6491326 |