Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stard13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163493.1 | NP_001156965.1 | 5 | 679 | ||
NM_146258.2 | NP_666370.3 | 5 | 679 | |||
GenBank mRNA | AK050259.1 | - | 5 | 680 | ||
BC027830.1 | - | 5 | 691 | 5320966 |