Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppm1k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175523.4 | NP_780732.1 | 3 | 731 | ||
GenBank mRNA | AK044610.1 | - | 3 | 801 | ||
BC092238.1 | - | 3 | 731 | 30642218 | ||
BC138473.1 | - | 2 | 537 | 8861493 | ||
BC138474.1 | - | 2 | 537 | 8861494 |