Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Leng9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175529.3 | NP_780738.1 | 1 | 1207 | ||
GenBank mRNA | AK089196.1 | - | 1 | 1207 | ||
AK090365.1 | - | 1 | 824 | |||
AK134559.1 | - | 1 | 828 | |||
AK147019.1 | - | 1709 | ||||
AK164490.1 | - | 1 | 1166 | |||
BC116676.1 | - | 1 | 1102 | 40106558 |