Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam71e2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172895.3 | NP_766483.3 | 9 | 1204 | ||
GenBank mRNA | AK076650.1 | - | 9 | 1262 |