Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028031.3 | NP_082307.1 | 9 | 1078 | ||
GenBank mRNA | AK010382.1 | - | 9 | 1078 | ||
AK049640.1 | - | 8 | 907 | |||
AK145855.1 | - | 9 | 998 | |||
AK146196.1 | - | 9 | 1050 | |||
BC019493.1 | - | 9 | 997 | |||
BC046599.1 | - | 9 | 997 | 6311721 |