Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081397.1 | NP_001074866.1 | 19 | 2493 | ||
GenBank mRNA | AB429291.1 | - | 16 | 1999 | ||
AK082350.1 | - | 19 | 2229 | |||
BC082311.1 | - | 19 | 2359 | |||
BC151049.1 | - | 19 | 2447 | 9087962 | ||
BC151051.1 | - | 19 | 2448 | 9087964 | ||
BC157966.1 | - | 19 | 2448 | 9007290 | ||
BC172122.1 | - | 19 | 2448 | 9054408 |