Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm4d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173433.2 | NP_775609.2 | 2 | 1983 | ||
GenBank mRNA | AK030020.1 | - | 2 | 1983 | ||
AK155227.1 | - | 3 | 2069 | |||
BC112372.1 | - | 1 | 986 |