Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp846 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172919.2 | NP_766507.2 | 6 | 1431 | ||
GenBank mRNA | AK008693.1 | - | 6 | 1265 | ||
AK033687.1 | - | 7 | 1337 | |||
BC139282.1 | - | 4 | 979 | 8862304 |