Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glb1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153803.1 | NP_722498.1 | 6 | 599 | ||
GenBank mRNA | AK033400.1 | - | 2 | 182 | ||
AK136711.1 | - | 6 | 633 | |||
BC038479.1 | - | 6 | 599 | 4503030 |