Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myrip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144557.5 | NP_653140.1 | 9 | 1077 | ||
GenBank mRNA | AB083782.1 | - | 8 | 916 | ||
AF396688.1 | - | 8 | 939 | |||
AK038988.1 | - | 9 | 1031 | |||
AK083225.1 | - | 9 | 1078 | |||
AK148358.1 | - | 9 | 1194 | |||
AY099470.1 | - | 8 | 952 | |||
BC129851.1 | - | 8 | 961 | 40109680 | ||
BC129852.1 | - | 8 | 961 | 40109682 |