Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gprasp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163015.1 | NP_001156487.1 | 7 | 3376 | ||
NM_001163016.1 | NP_001156488.1 | 7 | 3338 | |||
NM_001163017.1 | NP_001156489.1 | 7 | 3323 | |||
GenBank mRNA | AK030638.1 | - | 7 | 3316 | ||
BC117923.2 | - | 7 | 3342 | 40097504 | ||
BC117924.2 | - | 7 | 3292 | 40097505 |