Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amdhd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172935.4 | NP_766523.2 | 8 | 1021 | ||
GenBank mRNA | AK077584.1 | - | 8 | 1004 | ||
AK090009.1 | - | 8 | 1023 | |||
AK154209.1 | - | 8 | 1022 | |||
AK159860.1 | - | 8 | 1018 | |||
AK168432.1 | - | 8 | 979 | |||
AK179853.1 | - | 1 | 40 | |||
AK180149.1 | - | 1 | 40 | |||
AK180231.1 | - | 1 | 40 | |||
AK200617.1 | - | 3 | 340 | |||
BC037005.1 | - | 8 | 1057 | 5101453 |