Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hps5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005247.2 | NP_001005247.2 | 20 | 3365 | ||
NM_001005248.2 | NP_001005248.2 | 20 | 3341 | |||
NM_001167864.1 | NP_001161336.1 | 20 | 3266 | |||
GenBank mRNA | AF534396.1 | - | 20 | 3162 | ||
AF534397.1 | - | 20 | 3138 | |||
AF534398.1 | - | 18 | 2727 | |||
AF534399.1 | - | 21 | 3293 | |||
AK129265.1 | - | 20 | 3288 | |||
AK140780.1 | - | 9 | 1547 | |||
AK141088.1 | - | 19 | 3990 | |||
AK154612.1 | - | 20 | 3095 | |||
AK157182.1 | - | 20 | 3132 | |||
AK161039.1 | - | 11 | 1736 | |||
BC046405.1 | - | 15 | 2344 | |||
BC046443.1 | - | 20 | 2979 | |||
BC082542.1 | - | 20 | 3266 | 6405811 |