Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oas1h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159934.1 | NP_001153406.1 | 1 | 53 | ||
NM_145228.2 | NP_660263.1 | 1 | 53 | |||
GenBank mRNA | AB067530.1 | - | 1 | 53 | ||
AK135899.1 | - | 1 | 57 |