Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099738.1 | NP_001093208.1 | 2 | 933 | ||
NM_138664.2 | NP_619605.2 | 2 | 763 | |||
GenBank mRNA | AK081175.1 | - | 2 | 759 | ||
BC020175.1 | - | 2 | 757 | 4505154 |