Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc5a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133254.3 | NP_573517.1 | 12, 13 | 1661 | ||
GenBank mRNA | AK128900.1 | - | 12, 13 | 1675 | ||
AY033886.1 | - | 12, 13 | 1671 | |||
BC022226.1 | - | 12, 13 | 1661 | 4235707 |