Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1310 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146449.1 | NP_666660.1 | 1 | 111 | ||
GenBank mRNA | BC119359.1 | - | 1 | 137 | 8734108 |