Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr148 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146505.1 | NP_666716.1 | 1 | 430 | ||
GenBank mRNA | BC125353.1 | - | 1 | 455 | 40129868 | |
BC132239.1 | - | 1 | 455 | 40130516 |