Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1046 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146582.2 | NP_666793.2 | 1 | 457 | ||
GenBank mRNA | BC099891.1 | - | 1 | 456 | ||
BC099892.1 | - | 1 | 456 | |||
BC103540.1 | - | 1 | 456 |