Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr350 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146627.1 | NP_666838.1 | 1 | 919 | ||
GenBank mRNA | BC106790.1 | - | 1 | 919 | ||
BC106791.1 | - | 1 | 919 | |||
BC119406.2 | - | 1 | 919 | 40041609 |