Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr734 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146664.1 | NP_666875.1 | 1 | 777 | ||
GenBank mRNA | BC141029.1 | - | 1 | 812 | 9055641 |