Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1099 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146768.1 | NP_666979.1 | 1 | 464 | ||
GenBank mRNA | BC132348.1 | - | 1 | 486 | 40130625 | |
BC132350.1 | - | 1 | 486 | 40130627 |