Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153804.4 | NP_722499.4 | 2 | 499 | ||
GenBank mRNA | AK080928.1 | - | 2 | 500 | ||
AK162025.1 | - | 2 | 441 |