Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153804.4 | NP_722499.4 | 8 | 1245 | ||
GenBank mRNA | AK080928.1 | - | 8 | 1246 | ||
AK162025.1 | - | 8 | 1187 | |||
BC030417.1 | - | 7 | 588 | |||
BC049264.1 | - | 7 | 577 | |||
BC096443.1 | - | 8 | 1223 | 4189193 |