Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Angptl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011923.4 | NP_036053.2 | 2 | 637 | ||
GenBank mRNA | AF125176.1 | - | 1 | 124 | ||
AK011976.1 | - | 2 | 500 | |||
AK037265.1 | - | 2 | 530 | |||
AK155464.1 | - | 2 | 638 | |||
BC138609.1 | - | 2 | 443 | 8861629 | ||
BC138610.1 | - | 2 | 443 | 8861630 |