Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ceacam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039185.1 | NP_001034274.1 | 5 | 1130 | ||
NM_001039186.1 | NP_001034275.1 | 5 | 1130 | |||
NM_001039187.1 | NP_001034276.1 | 3 | 590 | |||
NM_011926.2 | NP_036056.2 | 3 | 590 | |||
GenBank mRNA | AB236328.2 | - | 3 | 499 | ||
AB236329.1 | - | 3 | 499 | |||
AB236330.2 | - | 4 | 784 | |||
AB236331.2 | - | 3 | 499 | |||
AB236332.2 | - | 5 | 1039 | |||
AK085408.1 | - | 713 | ||||
AK085749.1 | - | 5 | 1090 | |||
BC016891.1 | - | 5 | 1082 | |||
BC114998.1 | - | 1 | 62 | |||
BC121189.1 | - | 4 | 687 | |||
M61907.1 | - | 283 | ||||
M77196.1 | - | 5 | 1108 | |||
M96934.1 | - | 5 | 1056 | |||
M96935.1 | - | 4 | 492 | |||
X15351.1 | - | 5 | 1142 | |||
X53084.1 | - | 3 | 519 | |||
X67278.1 | - | 3 | 602 | |||
X67279.1 | - | 5 | 1142 | |||
X67280.1 | - | 5 | 1059 | |||
X67281.1 | - | 5 | 1059 | |||
X67282.1 | - | 3 | 602 | |||
X67283.1 | - | 3 | 519 |