Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ceacam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039185.1 | NP_001034274.1 | 5, 6 | 1331 | ||
NM_001039186.1 | NP_001034275.1 | 5, 6 | 1331 | |||
NM_001039187.1 | NP_001034276.1 | 3, 4 | 791 | |||
NM_011926.2 | NP_036056.2 | 3, 4 | 791 | |||
GenBank mRNA | AB236328.2 | - | 3, 4 | 700 | ||
AB236329.1 | - | 3, 4 | 700 | |||
AB236330.2 | - | 4, 5 | 985 | |||
BC016891.1 | - | 5, 6 | 1283 | |||
BC114998.1 | - | 1, 2 | 263 | |||
BC121189.1 | - | 4, 5 | 888 | |||
M61907.1 | - | 484 | ||||
M77196.1 | - | 5, 6 | 1309 | |||
M96934.1 | - | 5, 6 | 1257 | |||
M96935.1 | - | 4, 5 | 693 | |||
X15351.1 | - | 5, 6 | 1343 | |||
X53084.1 | - | 3, 4 | 720 | |||
X67278.1 | - | 3, 4 | 803 | |||
X67279.1 | - | 5, 6 | 1343 | |||
X67280.1 | - | 5, 6 | 1260 | |||
X67281.1 | - | 5, 6 | 1260 | |||
X67282.1 | - | 3, 4 | 803 | |||
X67283.1 | - | 3, 4 | 720 |