Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ceacam9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011927.4 | NP_036057.1 | 3 | 638 | ||
GenBank mRNA | AK049543.1 | - | 3 | 610 | ||
L49180.1 | - | 3 | 638 |