Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lypla2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011942.1 | NP_036072.1 | 5 | 297 | ||
GenBank mRNA | AB009653.1 | - | 5 | 297 | ||
AK003689.1 | - | 5 | 264 | |||
AK075590.1 | - | 5 | 289 | |||
AK089112.1 | - | 5 | 289 | |||
AK167478.1 | - | 5 | 284 | |||
BC068120.1 | - | 5 | 323 | 5714650 |