Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map3k3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011947.3 | NP_036077.1 | 16 | 1743 | ||
GenBank mRNA | AK137127.1 | - | 16 | 2029 | ||
AK220450.1 | - | 15 | 2633 | |||
BC023781.1 | - | 16 | 1747 | 5346982 | ||
U43187.1 | - | 16 | 2046 |