Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map3k4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011948.2 | NP_036078.2 | 4 | 1780 | ||
GenBank mRNA | AK122219.1 | - | 3 | 1463 | ||
AK133605.1 | - | 4 | 1917 | |||
BC058719.1 | - | 4 | 1780 | 5705378 | ||
U66240.1 | - | 2 | 663 | |||
U85607.1 | - | 4 | 1868 | |||
U85608.1 | - | 4 | 1868 |