Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map4k1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008279.2 | NP_032305.2 | 17, 18 | 1306 | ||
GenBank mRNA | AK143215.1 | - | 18, 19 | 1390 | ||
AK146273.1 | - | 17, 18 | 1290 | |||
BC005433.1 | - | 17, 18 | 1299 | 3494470 | ||
BC024832.1 | - | 4, 5 | 163 | |||
Y09010.1 | - | 17, 18 | 1342 |