Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map4k2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291787.1 | NP_001278716.1 | 27 | 2101 | ||
NM_009006.3 | NP_033032.1 | 27 | 2127 | |||
GenBank mRNA | AK011708.1 | - | 25 | 1992 | ||
AK042262.1 | - | 25 | 1849 | |||
AK051036.1 | - | 26 | 1916 | |||
AK078526.1 | - | 18 | 1355 | |||
AK154164.1 | - | 24 | 2572 | |||
AK154323.1 | - | 27 | 2032 | |||
BC034109.1 | - | 1 | 108 | |||
BC116286.1 | - | 26 | 2105 | 40094503 | ||
BC116287.1 | - | 27 | 1984 | 40094508 | ||
U50595.1 | - | 27 | 2037 |