Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038663.1 | NP_001033752.1 | 3 | 656 | ||
NM_011949.3 | NP_036079.1 | 3 | 656 | |||
GenBank mRNA | AK035386.1 | - | 3 | 630 | ||
AK048127.1 | - | 3 | 658 | |||
AK087925.1 | - | 3 | 631 | |||
AK132241.1 | - | 3 | 644 | |||
AK148853.1 | - | 3 | 636 | |||
AK218813.1 | - | 2 | 148 | |||
BB648734.1 | - | 3 | 657 | |||
BC058258.1 | - | 3 | 599 | 6468233 | ||
D10939.1 | - | 3 | 471 | |||
X58712.1 | - | 3 | 429 |