Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncdn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011986.4 | NP_036116.3 | 5 | 1986 | ||
GenBank mRNA | AK002938.1 | - | 3 | 591 | ||
AK122327.1 | - | 3 | 1331 | |||
AK149932.1 | - | 3 | 591 | |||
AK154889.1 | - | 5 | 1967 | |||
AK198940.1 | - | 1 | 53 | |||
BC017126.1 | - | 5 | 1917 | 4485839 | ||
BZ690994.2 | - | 1 | 24 | |||
CD353011.1 | - | 1 | 18 |