Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncdn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011986.4 | NP_036116.3 | 6 | 2049 | ||
GenBank mRNA | AK002938.1 | - | 4 | 654 | ||
AK122327.1 | - | 4 | 1394 | |||
AK154889.1 | - | 6 | 2030 | |||
AK170502.1 | - | 7 | 1764 | |||
BC017126.1 | - | 6 | 1980 | 4485839 | ||
BZ690994.2 | - | 2 | 87 | |||
CD353011.1 | - | 2 | 81 |