Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rcn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011992.2 | NP_036122.2 | 3 | 483 | ||
GenBank mRNA | AF049125.1 | - | 3 | 469 | ||
AK041996.1 | - | 480 | ||||
AK076153.1 | - | 3 | 310 | |||
AK077486.1 | - | 3 | 459 | |||
AK078106.1 | - | 3 | 451 | |||
AK169765.1 | - | 3 | 457 | |||
BB638743.1 | - | 3 | 480 | |||
BC132320.1 | - | 3 | 448 | 40130597 | ||
BC145668.1 | - | 3 | 448 | 40130971 |