Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277229.1 | NP_001264158.1 | 6 | 2249 | ||
NM_153398.3 | NP_700447.2 | 7 | 2315 | |||
GenBank mRNA | AK054097.1 | - | 7 | 2271 | ||
AK146986.1 | - | 7 | 2291 | |||
AK154467.1 | - | 7 | 2286 | |||
AY210402.1 | - | 7 | 2289 | |||
BC038139.1 | - | 1 | 564 | |||
BC050933.1 | - | 7 | 2277 | 6811382 | ||
BC055367.1 | - | 6 | 2192 | 30056072 |