Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Greb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252071.1 | NP_001239000.1 | 2 | 322 | ||
NM_015764.4 | NP_056579.2 | 4 | 644 | |||
GenBank mRNA | AF180470.1 | - | 3 | 300 | ||
AK045057.1 | - | 1 | 98 | |||
AK147337.1 | - | 2 | 172 | |||
AK157808.1 | - | 2 | 256 | |||
BC160931.1 | - | 3 | 254 | |||
CJ099922.1 | - | 4 | 443 |